Assay Details
Target Gene Details
Entrez Gene ID: | 67770 |
Gene Name: | caspase activity and apoptosis inhibitor 1 |
Gene Aliases: |
5830433M19Rik, AI536248, CAAP |
Location: |
Chr.4:94460277-94557076 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Caap1 | NM_026368.2 | 6 | 998 | NP_080644.2 |
AK076487.1 | 6 | 953 | ||
AK140382.1 | 6 | 999 | BAE24364.1 | |
AK150311.1 | 7 | 908 | ||
AK160297.1 | 6 | 1001 | ||
AK160572.1 | 6 | 999 | BAE35881.1 | |
AK171141.1 | 6 | 999 | ||
BC020067.1 | 6 | 986 | AAH20067.1 |
