Assay Details
Target Gene Details
Entrez Gene ID: | 12017 |
Gene Name: | BCL2-associated athanogene 1 |
Gene Aliases: |
BAG-1, Rap46 |
Location: |
Chr.4:40936398-40948294 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bag1 | NM_001171739.1 | 7 | 1214 | NP_001165210.1 |
NM_009736.3 | 7 | 1214 | NP_033866.4 | |
AF022223.1 | 7 | 1155 | AAC34259.1 | |
AK009149.1 | 7 | 1125 | BAB26106.1 | |
AK013611.1 | 5 | 500 | BAB28930.1 | |
AK154469.1 | 7 | 1163 | ||
AK171094.1 | 7 | 1163 | ||
AK184952.1 | 1 | 90 | ||
AK213721.1 | 1 | 90 | ||
AK216811.1 | 1 | 90 | ||
BC003722.1 | 7 | 1176 | ||
BC069918.1 | 7 | 1151 | ||
BC093509.1 | 7 | 1125 | ||
BQ031093.1 | 1 | 152 |
