Assay Details
Target Gene Details
Entrez Gene ID: | 654409 |
Gene Name: | RIKEN cDNA 4932416H05 gene |
Gene Aliases: |
- |
Location: |
Chr.2:129798369-129801459 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4932416H05Rik | NR_029452.1 | 1 | 892 | |
| AB041802.1 | 1 | 878 | ||
| AK016906.1 | 1 | 888 | ||
| AK030036.1 | 1 | 893 |
Target Gene Details
Entrez Gene ID: | 67333 |
Gene Name: | serine/threonine kinase 35 |
Gene Aliases: |
1700054C12Rik, CLIK1, Stk35l1 |
Location: |
Chr.2:129800517-129832287 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk35 | NM_001038635.2 | 1 | 52 | NP_001033724.2 |
| NM_183262.3 | 1 | 52 | NP_899085.3 | |
| BC138904.1 | ||||
| BC138905.1 | ||||
| CJ073113.1 | 1 | 56 | ||
| CJ113079.1 |
More Information
Set Membership: |
|

Genomic Map