Assay Details
Target Gene Details
Entrez Gene ID: | 53416 |
Gene Name: | serine/threonine kinase 39 |
Gene Aliases: |
AW227544, AW556857, DCHT, RF005, Rnl5, SPAK |
Location: |
Chr.2:68210445-68472965 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk39 | NM_016866.2 | 1 | 55 | NP_058562.1 |
| AF099988.1 | AAC72237.1 | |||
| AK017292.1 | ||||
| AK148456.1 | 1 | 91 | ||
| BC051964.1 | 1 | 342 | AAH51964.2 | |
| BC064443.1 | 1 | 55 | AAH64443.1 |

Genomic Map