Assay Details
Target Gene Details
Entrez Gene ID: | 229541 |
Gene Name: | DENN/MADD domain containing 4B |
Gene Aliases: |
- |
Location: |
Chr.3:90264920-90280665 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd4b | NM_201407.4 | 3 | 578 | NP_958809.3 |
| AK155638.1 | 3 | 560 | BAE33359.1 | |
| AY512564.1 | 2 | 377 | AAR97575.1 | |
| BC066102.1 | 2 | 357 | ||
| BC151194.1 | 2 | 224 |

Genomic Map