Assay Details
Target Gene Details
Entrez Gene ID: | 67553 |
Gene Name: | glutathione S-transferase, C-terminal domain containing |
Gene Aliases: |
4933434L15Rik |
Location: |
Chr.3:132981752-133091971 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gstcd | NM_026231.2 | 3 | 689 | NP_080507.2 |
AK017059.1 | BAB30574.2 | |||
AK020021.1 | 2 | 299 | BAB31969.1 | |
AK029724.1 | 3 | 688 | ||
AK033409.1 | 3 | 727 | ||
AK037169.1 | 3 | 728 | BAC29731.1 | |
AK044441.1 | 3 | 697 | BAC31921.1 | |
BC047136.1 | 3 | 689 | AAH47136.1 | |
BC061240.1 | 3 | 685 | AAH61240.1 |
