Assay Details
Target Gene Details
Entrez Gene ID: | 68576 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 5 |
Gene Aliases: |
1110003H18Rik, Hbxip, XIP |
Location: |
Chr.3:107278858-107284082 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamtor5 | NM_026774.2 | 1 | 165 | NP_081050.2 |
AI552736.1 | 1 | 165 | ||
AK003358.1 | BAB22734.1 | |||
AK146289.1 | BAE27046.1 | |||
BC028547.1 | AAH28547.1 |
