Assay Details
Target Gene Details
Entrez Gene ID: | 242235 |
Gene Name: | leucine-rich repeat, immunoglobulin-like and transmembrane domains 3 |
Gene Aliases: |
Gm421 |
Location: |
Chr.3:129786929-129808884 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrit3 | NM_001287224.1 | 4 | 1664 | NP_001274153.1 |
| KF954709.1 | 4 | 1664 |
More Information
Set Membership: |
|

Genomic Map