Assay Details
Target Gene Details
Entrez Gene ID: | 11532 |
Gene Name: | alcohol dehydrogenase 5 (class III), chi polypeptide |
Gene Aliases: |
Adh-5, Adh3, GSNOR |
Location: |
Chr.3:138437200-138455499 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adh5 | NM_001288578.1 | 6 | 810 | NP_001275507.1 |
| NM_007410.3 | 6 | 828 | NP_031436.2 | |
| AK002790.1 | 6 | 750 | ||
| AK076507.1 | 6 | 757 | BAC36370.1 | |
| AK146949.1 | 6 | 772 | BAE27558.1 | |
| AK159803.1 | 6 | 808 | BAE35383.1 | |
| AK214149.1 | 2 | 286 | ||
| BC062879.1 | 6 | 754 | ||
| BC090978.1 | 6 | 751 | AAH90978.1 | |
| M84147.1 | 6 | 778 | AAA68896.1 |
More Information
Set Membership: |
|

Genomic Map