Assay Details
Target Gene Details
Entrez Gene ID: | 66536 |
Gene Name: | nipsnap homolog 3B (C. elegans) |
Gene Aliases: |
2700063N13Rik, Nipsnap3a |
Location: |
Chr.4:53011775-53022060 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nipsnap3b | NM_025623.2 | 6 | 979 | NP_079899.1 |
| AF353623.1 | 6 | 968 | AAL56225.1 | |
| AK004458.1 | 6 | 978 | BAB23312.1 | |
| AK008786.1 | 6 | 975 | BAB25894.1 | |
| AK012480.1 | 6 | 979 | BAB28269.1 | |
| AK080363.1 | ||||
| BC019505.1 | 6 | 964 | AAH19505.1 |

Genomic Map