Assay Details
Target Gene Details
Entrez Gene ID: | 319974 |
Gene Name: | autism susceptibility candidate 2 |
Gene Aliases: |
2700063G02Rik, A730011F23Rik, D830032G16Rik |
Location: |
Chr.5:131437682-132542343 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Auts2 | NM_177047.3 | 19 | 5556 | NP_796021.2 |
| AK028848.1 | 1 | 2093 | BAC26152.1 | |
| AK035818.1 | 1 | 2145 | BAC29198.1 | |
| AK135954.1 | 1 | 620 | ||
| AK164158.1 | 15 | 4996 | ||
| BC058110.1 | 7 | 3132 | AAH58110.1 | |
| CA320283.1 | 1 | 235 |

Genomic Map