Assay Details
Target Gene Details
Entrez Gene ID: | 631037 |
Gene Name: | cytochrome P450, family 4, subfamily b, polypeptide 1, pseudogene 2 |
Gene Aliases: |
Gm12839, OTTMUSG00000008630 |
Location: |
Chr.4:115582038-115583127 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp4b1-ps2 | NR_033575.1 | 3 | 455 | |
AA185606.1 | 3 | 455 |
