Assay Details
Target Gene Details
Entrez Gene ID: | 71514 |
Gene Name: | splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated) |
Gene Aliases: |
1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1 |
Location: |
Chr.4:127020560-127037016 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sfpq | NM_023603.3 | 1 | 767 | NP_076092.1 |
| NR_045010.1 | 1 | 767 | ||
| AF272847.1 | 1 | 60 | AAG17365.1 | |
| AK157783.1 | 2 | 404 | BAE34196.1 | |
| AY034062.1 | 1 | 682 | AAK60397.1 | |
| BC089305.1 | 1 | 733 | AAH89305.1 |
More Information
Set Membership: |
|

Genomic Map