Assay Details
Target Gene Details
Entrez Gene ID: | 545681 |
Gene Name: | predicted gene 12992 |
Gene Aliases: |
OTTMUSG00000009742 |
Location: |
Chr.4:131899478-131920029 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm12992 | NR_102393.1 | 5 | 3466 | |
| AK047558.1 | 4 | 3204 |
Target Gene Details
Entrez Gene ID: | 57317 |
Gene Name: | serine/arginine-rich splicing factor 4 |
Gene Aliases: |
5730499P16Rik, AW550192, MNCb-2616, SRp75, Sfrs4 |
Location: |
Chr.4:131873611-131901725 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf4 | NM_020587.2 | 6 | 1034 | NP_065612.2 |
| AB041587.1 | 3 | 472 | BAA95070.1 | |
| AK075769.1 | 6 | 1002 | ||
| AK077648.1 | 6 | 1020 | BAC36925.1 | |
| AK156989.1 | 6 | 969 | BAE33925.1 | |
| BC019437.1 | 6 | 994 | AAH19437.1 | |
| BC026944.1 | 6 | 1900 | AAH26944.1 |
More Information
Set Membership: |
|

Genomic Map