Assay Details
Target Gene Details
Entrez Gene ID: | 117197 |
Gene Name: | biogenesis of lysosomal organelles complex-1, subunit 4, cappuccino |
Gene Aliases: |
2610101N07Rik, Blos4, Cno |
Location: |
Chr.5:36747374-36748679 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bloc1s4 | NM_133724.3 | 1 | 870 | NP_598485.1 |
| AK011795.1 | 1 | 843 | ||
| AK182170.1 | ||||
| AK207225.1 | 1 | 103 | ||
| AK207410.1 | 1 | 103 | ||
| BC019169.1 | 1 | 860 | AAH19169.1 |

Genomic Map