Assay Details
Target Gene Details
Entrez Gene ID: | 67305 |
Gene Name: | glutathione peroxidase 7 |
Gene Aliases: |
3110050F08Rik, AI327032, GPX6 |
Location: |
Chr.4:108400217-108406713 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx7 | NM_024198.3 | 2 | 345 | NP_077160.1 |
| AK004091.1 | 2 | 338 | BAB23164.1 | |
| AK014199.1 | 2 | 343 | ||
| AK084382.1 | 2 | 346 | ||
| AK165221.1 | 2 | 346 | BAE38085.1 | |
| BC003228.1 | 2 | 346 | AAH03228.1 |
More Information
Set Membership: |
|

Genomic Map