Assay Details
Target Gene Details
Entrez Gene ID: | 22385 |
Gene Name: | bromodomain adjacent to zinc finger domain, 1B |
Gene Aliases: |
C87820, WSTF, Wbscr9 |
Location: |
Chr.5:135187323-135246129 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz1b | NM_011714.2 | 1 | 364 | NP_035844.2 |
AF084480.1 | 1 | 325 | AAD08676.1 | |
AK017894.1 | 1 | 365 | BAB30992.1 | |
AK140172.1 | 1 | 152 | BAE24264.1 | |
AK141305.1 | 1 | 105 | BAE24643.1 | |
BC141399.1 | 1 | 267 |
