Assay Details
Target Gene Details
Entrez Gene ID: | 74610 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 8 |
Gene Aliases: |
4833412N02Rik, AA409895 |
Location: |
Chr.5:24393681-24409947 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb8 | NM_029020.2 | 16 | 2557 | NP_083296.2 |
| AF213391.1 | 4 | 572 | AAF31430.1 | |
| AK014319.1 | 16 | 2288 | BAB29270.1 | |
| AK030624.1 | 16 | 2284 | BAC27052.1 | |
| AK038760.1 | 16 | 2285 | ||
| AK049152.1 | 16 | 2301 | BAC33571.1 | |
| AK076315.1 | 16 | 2308 | BAC36297.1 | |
| AK166704.1 | 16 | 2291 | BAE38958.1 | |
| AK170334.1 | 4 | 577 | ||
| AK178570.1 | 2 | 267 | ||
| AK185108.1 | 1 | 150 | ||
| AK186503.1 | ||||
| AK186540.1 | 1 | 150 | ||
| AK208007.1 | 1 | 150 | ||
| BC015301.1 | 16 | 2557 | AAH15301.1 |
Target Gene Details
Entrez Gene ID: | 12568 |
Gene Name: | cyclin-dependent kinase 5 |
Gene Aliases: |
AW048668, Crk6 |
Location: |
Chr.5:24407597-24423591 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5 |
More Information
Set Membership: |
|

Genomic Map