Assay Details
Target Gene Details
Entrez Gene ID: | 381680 |
Gene Name: | neurexophilin and PC-esterase domain family, member 5 |
Gene Aliases: |
- |
Location: |
Chr.5:138207499-138251876 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nxpe5 | NM_001013773.3 | 2 | 170 | NP_001013795.3 |
| AK154713.1 | 2 | 178 | BAE32781.1 | |
| AK156114.1 | 2 | 172 | ||
| AK171365.1 | 2 | 178 | BAE42413.1 |

Genomic Map