Assay Details
Target Gene Details
Entrez Gene ID: | 68817 |
Gene Name: | DNA-damage inducible protein 2 |
Gene Aliases: |
1110056G13Rik, 1700027M01Rik, 9130022E05Rik, AI604911, AU040698 |
Location: |
Chr.4:141683563-141723419 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddi2 | NM_001017966.2 | 10 | 3199 | NP_001017966.1 |
Target Gene Details
Entrez Gene ID: | 69994 |
Gene Name: | regulatory solute carrier protein, family 1, member 1 |
Gene Aliases: |
1700027M01Rik, OTTMUSG00000010365, Rs1 |
Location: |
Chr.4:141683563-141685716 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsc1a1 | NM_023544.5 | 1 | 1814 | NP_076033.4 |
| AK006435.1 | 1 | 628 | BAB24588.1 | |
| BC150752.1 | 1 | 1814 |
More Information
Set Membership: |
|

Genomic Map