Assay Details
Target Gene Details
Entrez Gene ID: | 13200 |
Gene Name: | dolichyl-di-phosphooligosaccharide-protein glycotransferase |
Gene Aliases: |
- |
Location: |
Chr.4:138304738-138312611 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddost | NM_007838.2 | 11 | 1707 | NP_031864.2 |
AK077651.1 | 11 | 1569 | ||
AK081542.1 | BAC38250.1 | |||
AK084267.1 | 11 | 1592 | ||
AK148095.1 | BAE28341.1 | |||
AK150683.1 | BAE29763.1 | |||
AK190281.1 | ||||
AK191754.1 | ||||
BC068132.1 | 11 | 1707 | AAH68132.1 | |
D89063.1 | BAA23671.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
