Assay Details
Target Gene Details
Entrez Gene ID: | 230073 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 58 |
Gene Aliases: |
6430573D20Rik, C330021E21, RIG-I, RLR-1 |
Location: |
Chr.4:40203777-40239825 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx58 | NM_172689.3 | 8 | 1173 | NP_766277.3 |
AK052595.1 | 4 | 427 | ||
AK052871.1 | 8 | 1158 | BAC35183.1 | |
AK078287.1 | 8 | 1105 | BAC37205.1 | |
AK128929.1 | 8 | 1108 | ||
AK155930.1 | 8 | 1109 | BAE33508.1 | |
AY553221.1 | 8 | 1049 | AAS59532.1 | |
BC027369.1 | 8 | 1152 | ||
BC106095.1 | 8 | 1093 | AAI06096.1 | |
BC127163.1 | 8 | 1039 | ||
BC152540.1 | 8 | 1041 |
