Assay Details
Target Gene Details
Entrez Gene ID: | 11409 |
Gene Name: | acyl-Coenzyme A dehydrogenase, short chain |
Gene Aliases: |
AI196007, Bcd-1, Bcd1, Hdlq8, SCAD |
Location: |
Chr.5:115110299-115119346 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acads | NM_007383.3 | 10 | 1749 | NP_031409.2 |
| AK155361.1 | 10 | 1728 | BAE33217.1 | |
| AK169428.1 | 10 | 1686 | BAE41170.1 | |
| BC016259.1 | 10 | 1716 | AAH16259.1 | |
| BE993251.1 | 1 | 133 | ||
| L11163.1 | 10 | 1673 | AAA16714.1 |
More Information
Set Membership: |
|

Genomic Map