Assay Details
Target Gene Details
Entrez Gene ID: | 230098 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 39 |
Gene Aliases: |
E130306D19Rik |
Location: |
Chr.4:43496139-43508769 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef39 |
Target Gene Details
Entrez Gene ID: | 230099 |
Gene Name: | carbonic anhydrase 9 |
Gene Aliases: |
CAIX, Ca9, MN/CA9 |
Location: |
Chr.4:43507026-43513810 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car9 | NM_139305.2 | 1 | 302 | NP_647466.2 |
| AB086322.1 | 1 | 304 | BAC00816.1 | |
| AJ245857.1 | 1 | 304 | CAC80975.1 | |
| AK136579.1 | 1 | 303 | BAE23060.1 | |
| AK137719.1 | 1 | 304 | BAE23474.1 | |
| BC120544.1 | 1 | 295 | ||
| BC120546.1 | 1 | 295 | ||
| EF122497.1 | 1 | 304 |

Genomic Map