Assay Details
Target Gene Details
Entrez Gene ID: | 15444 |
Gene Name: | hippocalcin |
Gene Aliases: |
- |
Location: |
Chr.4:129111570-129122070 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hpca | NM_001130419.2 | 4 | 1160 | NP_001123891.1 |
| NM_001286081.1 | 4 | 1073 | NP_001273010.1 | |
| NM_001286083.1 | 4 | 1005 | NP_001273012.1 | |
| NM_010471.4 | 4 | 1064 | NP_034601.1 | |
| AK002992.1 | 4 | 1064 | BAB22502.1 | |
| BC049607.1 | 4 | 1029 | AAH49607.1 | |
| BC058588.1 | 3 | 978 | AAH58588.1 |

Genomic Map