Assay Details
Target Gene Details
Entrez Gene ID: | 72098 |
Gene Name: | transmembrane protein 68 |
Gene Aliases: |
2010300G19Rik, AA408325, AW123402 |
Location: |
Chr.4:3549041-3574848 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem68 | NM_028097.3 | 8 | 2096 | NP_082373.1 |
AK008493.1 | 8 | 2130 | BAB25698.1 | |
AK048210.1 | 8 | 2096 | BAC33273.1 | |
AK084881.1 | 8 | 2130 | BAC39301.1 | |
AK088600.1 | 9 | 2266 | BAC40445.1 | |
AK147765.1 | 8 | 2129 | BAE28123.1 | |
AK162563.1 | 8 | 2129 | BAE36969.1 | |
AK165916.1 | 8 | 2158 | BAE38459.1 | |
AK166237.1 | 8 | 2182 | BAE38652.1 | |
BC016240.1 | 8 | 2114 | AAH16240.1 |
