Assay Details
Target Gene Details
Entrez Gene ID: | 319885 |
Gene Name: | zinc finger, CCHC domain containing 7 |
Gene Aliases: |
4930572I07Rik, D4Wsu132e |
Location: |
Chr.4:44756559-44932215 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zcchc7 | NM_138590.4 | 9 | 2316 | NP_613056.2 |
AA277854.1 | 1 | 238 | ||
AK013088.1 | 1 | 846 | ||
AK013175.1 | 3 | 1800 | BAB28692.1 | |
AK050430.1 | 3 | 3008 | BAC34252.1 | |
BC054405.1 | 3 | 1519 | AAH54405.1 |
