Assay Details
Target Gene Details
Entrez Gene ID: | 319885 |
Gene Name: | zinc finger, CCHC domain containing 7 |
Gene Aliases: |
4930572I07Rik, D4Wsu132e |
Location: |
Chr.4:44756559-44932215 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zcchc7 | NM_138590.4 | 2 | 338 | NP_613056.2 |
AK029808.1 | 2 | 345 | ||
AK032353.1 | 2 | 310 | ||
AK039158.1 | 2 | 259 | ||
AK043238.1 | 2 | 251 | BAC31499.1 | |
AK080785.1 | 2 | 251 | ||
AK157728.1 | 2 | 241 | BAE34173.1 | |
BC117020.1 | 2 | 241 | ||
BC117022.1 | 2 | 241 | ||
BC144774.1 | 2 | 245 | ||
CJ150974.1 | ||||
CJ155194.1 | 2 | 310 |
