Assay Details
Target Gene Details
Entrez Gene ID: | 329872 |
Gene Name: | Fras1 related extracellular matrix protein 1 |
Gene Aliases: |
BC037594, D430009N09, D630008K06, crf11, eye<m02Jus>, eyes2, heb, qbrick |
Location: |
Chr.4:82897920-83052548 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Frem1 | NM_001198811.1 | 16 | 3350 | NP_001185740.1 |
NM_177863.4 | 15 | 3140 | NP_808531.2 | |
AB160986.1 | 16 | 3350 | BAD89014.1 | |
AJ616838.1 | 15 | 3140 | CAE83577.1 | |
BC138427.1 | 15 | 3051 |
