Assay Details
Target Gene Details
Entrez Gene ID: | 19359 |
Gene Name: | RAD23b homolog (S. cerevisiae) |
Gene Aliases: |
0610007D13Rik, AV001138, HR23B, mHR23B, p58 |
Location: |
Chr.4:55350042-55392237 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad23b | NM_009011.4 | 10 | 3423 | NP_033037.2 |
| AK142280.1 | 10 | 3400 | BAE25006.1 | |
| BE989853.1 |
More Information
Set Membership: |
|

Genomic Map