Assay Details
Target Gene Details
Entrez Gene ID: | 19359 |
Gene Name: | RAD23b homolog (S. cerevisiae) |
Gene Aliases: |
0610007D13Rik, AV001138, HR23B, mHR23B, p58 |
Location: |
Chr.4:55350042-55392237 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad23b | NM_009011.4 | 4 | 770 | NP_033037.2 |
| AK142280.1 | 4 | 762 | BAE25006.1 | |
| AK150089.1 | 4 | 762 | BAE29298.1 | |
| AK157252.1 | 4 | 732 | BAE34014.1 | |
| AK160880.1 | 4 | 762 | BAE36067.1 | |
| AK160890.1 | 4 | 748 | BAE36071.1 | |
| AK160973.1 | 4 | 748 | BAE36124.1 | |
| AK167582.1 | 4 | 758 | BAE39643.1 | |
| AK192062.1 | 1 | 131 | ||
| AK200932.1 | 1 | 40 | ||
| BC027747.1 | 4 | 759 | AAH27747.1 | |
| BC068193.1 | 4 | 729 | AAH68193.1 | |
| X92411.1 | 4 | 450 | CAA63146.1 |
More Information
Set Membership: |
|

Genomic Map