Assay Details
Target Gene Details
Entrez Gene ID: | 242291 |
Gene Name: | inositol monophosphatase domain containing 1 |
Gene Aliases: |
1110001C20Rik, AA408880, AI451589, AL022796, B230207P20, Jaws, gPAPP |
Location: |
Chr.4:4764342-4793306 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Impad1 | NM_177730.3 | 5 | 2827 | NP_808398.1 |
| AK045511.1 | 5 | 2877 | ||
| BC048776.1 | 5 | 2827 | AAH48776.1 | |
| BC138209.1 | 5 | 2684 | ||
| BC145952.1 | 5 | 2684 |
More Information
Set Membership: |
|

Genomic Map