Assay Details
Target Gene Details
Entrez Gene ID: | 214944 |
Gene Name: | MOB kinase activator 3B |
Gene Aliases: |
8430436F23Rik, A430018A01Rik, Mobkl2b |
Location: |
Chr.4:34949074-35157495 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob3b | NM_178061.5 | 2 | 446 | NP_835162.1 |
| AK039841.1 | 2 | 292 | BAC30466.1 | |
| AK087285.1 | 2 | 447 | ||
| AK155217.1 | 2 | 447 | BAE33128.1 | |
| AK170973.1 | 2 | 440 | BAE42152.1 | |
| BC020028.1 | 2 | 428 | AAH20028.1 |
More Information
Set Membership: |
|

Genomic Map