Assay Details
Target Gene Details
Entrez Gene ID: | 66096 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 4 |
Gene Aliases: |
0910001L09Rik, AV006840 |
Location: |
Chr.5:138255482-138259395 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamtor4 | NM_001081108.2 | 1 | 267 | NP_001074577.1 |
| AK002488.1 | BAC24993.1 | |||
| AK003086.1 | BAC25020.1 | |||
| AK003250.1 | ||||
| AK010598.1 | BAC25306.1 | |||
| AK146994.1 | ||||
| BC144958.1 | 1 | 267 |

Genomic Map