Assay Details
Target Gene Details
Entrez Gene ID: | 14380 |
Gene Name: | glucose-6-phosphate dehydrogenase 2 |
Gene Aliases: |
G6pdx-ps1, Gpd-2, Gpd2 |
Location: |
Chr.5:61808843-61810477 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G6pd2 | NM_019468.2 | 1 | 537 | NP_062341.2 |
| BC120827.1 | 1 | 537 | ||
| BC137684.1 | 1 | 537 |
More Information
Set Membership: |
|

Genomic Map