Assay Details
Target Gene Details
Entrez Gene ID: | 12579 |
Gene Name: | cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4) |
Gene Aliases: |
AV083695, INK4b, MTS2, p15, p15(INK4b), p15INK4b |
Location: |
Chr.4:89306289-89311032 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn2b | NM_007670.4 | 2 | 1223 | NP_031696.1 |
| AF059567.1 | 2 | 1132 | AAC14569.1 | |
| AK144132.1 | 2 | 1233 | BAE25719.1 | |
| AK146668.1 | ||||
| BC002010.1 | 2 | 1137 | AAH02010.1 |

Genomic Map