Assay Details
Target Gene Details
Entrez Gene ID: | 14610 |
Gene Name: | gap junction protein, alpha 10 |
Gene Aliases: |
Cx-57, Cx59, cx57 |
Location: |
Chr.4:32600865-32602382 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gja10 | NM_010289.2 | 1 | 759 | NP_034419.2 |
More Information
Set Membership: |
|

Genomic Map