Assay Details
Target Gene Details
Entrez Gene ID: | 50780 |
Gene Name: | regulator of G-protein signaling 3 |
Gene Aliases: |
4930506N09Rik, C2PA-RGS3, C2pa, PDZ-RGS3, RGS3S |
Location: |
Chr.4:62559838-62703022 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs3 | NM_001081650.2 | 11 | 3543 | NP_001075119.1 |
NM_019492.3 | 20 | 4247 | NP_062365.2 | |
NM_134257.3 | NP_599018.3 | |||
AF319519.1 | 11 | 3556 | AAL37173.1 | |
AF350047.1 | AAK38878.1 | |||
AJ250999.1 | 20 | 4247 | CAB66146.1 | |
AK004648.1 | BAB23439.1 | |||
AK028445.1 | 11 | 3528 | BAC25954.1 | |
AK041392.1 | ||||
AK046265.1 | 2 | 2556 | ||
AK136567.1 | 1 | 2198 | BAE23049.1 | |
BY704855.1 |
