Assay Details
Target Gene Details
Entrez Gene ID: | 50780 |
Gene Name: | regulator of G-protein signaling 3 |
Gene Aliases: |
4930506N09Rik, C2PA-RGS3, C2pa, PDZ-RGS3, RGS3S |
Location: |
Chr.4:62559838-62703022 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs3 | NM_001081650.2 | 4 | 482 | NP_001075119.1 |
NM_019492.3 | 13 | 1186 | NP_062365.2 | |
NM_134257.3 | 4 | 482 | NP_599018.3 | |
AF319519.1 | 4 | 495 | AAL37173.1 | |
AF350047.1 | 4 | 465 | AAK38878.1 | |
AJ250999.1 | 13 | 1186 | CAB66146.1 | |
AK015722.1 | 11 | 941 | ||
AK028445.1 | 4 | 467 | BAC25954.1 | |
BB653029.1 |
