Assay Details
Target Gene Details
Entrez Gene ID: | 68955 |
Gene Name: | serine/arginine repetitive matrix 4 |
Gene Aliases: |
1500001A10Rik, B230202K19Rik, bv, mKIAA1853, nSR100 |
Location: |
Chr.5:116439272-116592562 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srrm4 | NM_026886.3 | 13 | 2814 | NP_081162.1 |
| AK053836.1 | 13 | 2546 | BAC35548.1 | |
| AK129457.1 | 5 | 1896 | BAC98267.1 | |
| BC024130.1 | 2 | 1296 | AAH24130.1 | |
| BC056392.1 | 13 | 2804 | AAH56392.1 |
Target Gene Details
Entrez Gene ID: | 76798 |
Gene Name: | serine/arginine repetitive matrix 4, opposite strand |
Gene Aliases: |
2410137F16Rik |
Location: |
Chr.5:116438722-116465487 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srrm4os | NR_015595.2 | |||
| AK010799.1 | ||||
| AK049160.1 | ||||
| AK082818.1 |
More Information
Set Membership: |
|

Genomic Map