Assay Details
Target Gene Details
Entrez Gene ID: | 12293 |
Gene Name: | calcium channel, voltage-dependent, alpha2/delta subunit 1 |
Gene Aliases: |
Ca(v)alpha2delta1, Cacna2, Cchl2a |
Location: |
Chr.5:15934691-16374511 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna2d1 | NM_001110843.1 | 1 | 345 | NP_001104313.1 |
| NM_001110844.1 | 1 | 345 | NP_001104314.1 | |
| NM_001110845.1 | 1 | 345 | NP_001104315.1 | |
| NM_001110846.1 | 1 | 345 | NP_001104316.1 | |
| NM_009784.2 | 1 | 345 | NP_033914.1 | |
| AK052898.1 | 1 | 125 | BAC35195.1 | |
| AK084101.1 | 1 | 298 | ||
| AK164580.1 | 1 | 346 | ||
| BC115871.1 | 2 | 120 | ||
| U73483.1 | 1 | 256 | AAB50138.1 | |
| U73484.1 | 1 | 256 | AAB50139.1 | |
| U73485.1 | 1 | 256 | AAB50140.1 | |
| U73486.1 | 1 | 256 | AAB50141.1 | |
| U73487.1 | 1 | 256 | AAB50142.1 |
More Information
Set Membership: |
|

Genomic Map