Assay Details
Target Gene Details
Entrez Gene ID: | 140904 |
Gene Name: | calneuron 1 |
Gene Aliases: |
9630012C17Rik, Cabp8, MNCb-0849 |
Location: |
Chr.5:130369356-130846325 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Caln1 | NM_021371.2 | 1 | 221 | NP_067346.1 |
| NM_181045.1 | NP_851388.1 | |||
| AB041539.1 | 1 | 212 | BAA95024.1 | |
| AF282251.1 | AAK15156.1 | |||
| AK035865.1 | BAC29219.1 | |||
| AK081011.1 | BAC38115.1 | |||
| AK158892.1 | 1 | 193 | BAE34716.1 | |
| BC057129.1 | 1 | 200 | AAH57129.1 | |
| BC058963.1 | 1 | 200 | AAH58963.1 | |
| BY274079.1 | 1 | 221 |

Genomic Map