Assay Details
Target Gene Details
Entrez Gene ID: | 79554 |
Gene Name: | ceramide-1-phosphate transfer protein |
Gene Aliases: |
Gltpd1 |
Location: |
Chr.4:155864723-155869440 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cptp | NM_024472.4 | 1 | 230 | NP_077792.2 |
| AK040664.1 | 1 | 231 | BAC30659.1 | |
| AK089229.1 | 1 | 200 |
More Information
Set Membership: |
|

Genomic Map