Assay Details
Target Gene Details
Entrez Gene ID: | 29864 |
Gene Name: | ring finger protein 11 |
Gene Aliases: |
- |
Location: |
Chr.4:109452473-109484723 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf11 | NM_013876.3 | 3 | 1345 | NP_038904.1 |
| AB024427.1 | 4 | 1210 | BAA84682.1 | |
| AK014254.1 | 3 | 1499 | ||
| BC010299.1 | 3 | 1345 | AAH10299.1 |

Genomic Map