Assay Details
Target Gene Details
Entrez Gene ID: | 13486 |
Gene Name: | down-regulator of transcription 1 |
Gene Aliases: |
1700121L09Rik, Dr1l, NC2, NC2beta |
Location: |
Chr.5:108268897-108280521 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dr1 | NM_026106.4 | 3 | 1414 | NP_080382.2 |
| AK007230.1 | 3 | 1389 | ||
| AK018040.1 | 3 | 1337 | ||
| AK031057.1 | 3 | 1348 | BAE20474.1 | |
| AK075713.1 | 3 | 1414 | BAC35903.1 | |
| AK090113.1 | 3 | 1396 | BAC41099.1 | |
| AK139866.1 | 3 | 1344 | BAE24166.1 | |
| AK144749.1 | 3 | 1408 | BAE26046.1 | |
| AK146573.1 | 3 | 1377 | BAE27269.1 | |
| BC013461.1 | 3 | 983 | AAH13461.1 |

Genomic Map