Assay Details
Target Gene Details
Entrez Gene ID: | 118446 |
Gene Name: | gap junction protein, gamma 3 |
Gene Aliases: |
Cx29, Gje1 |
Location: |
Chr.5:137953809-137962959 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjc3 | NM_080450.4 | 2 | 358 | NP_536698.2 |
| AK034037.1 | 2 | 323 | BAC28554.1 | |
| AK045577.1 | 2 | 498 | BAC32421.1 | |
| AK047368.1 | 2 | 359 | BAC33038.1 | |
| AK147913.1 | 2 | 498 | BAE28224.1 | |
| AY289210.1 | 1 | 263 | AAP41202.1 | |
| AY293564.1 | 1 | 263 | AAP50246.1 | |
| AY297108.1 | 1 | 263 | AAP51160.1 | |
| BC125305.1 | 2 | 345 |
More Information
Set Membership: |
|

Genomic Map