Assay Details
Target Gene Details
Entrez Gene ID: | 75814 |
Gene Name: | RIKEN cDNA 4930467D21 gene |
Gene Aliases: |
4930448L09Rik, BB013444 |
Location: |
Chr.5:97392417-97588126 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930467D21Rik | NR_045981.1 | |||
| AK015518.1 | ||||
| AK133367.1 |
Target Gene Details
Entrez Gene ID: | 14626 |
Gene Name: | glycerol kinase 2 |
Gene Aliases: |
GKTA, Gk-rs2 |
Location: |
Chr.5:97455161-97457008 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gk2 | NM_010294.1 | 1 | 1550 | NP_034424.1 |
| AF117734.1 | 1 | 1550 | AAD24551.1 | |
| AK075761.1 | 1 | 1463 | BAC35939.1 | |
| BC050764.1 | 1 | 1563 | AAH50764.1 | |
| BC061147.1 | 1 | 1529 | AAH61147.1 |
More Information
Set Membership: |
|

Genomic Map