Assay Details
Target Gene Details
Entrez Gene ID: | 19934 |
Gene Name: | ribosomal protein L22 |
Gene Aliases: |
2700038K18Rik |
Location: |
Chr.4:152324436-152334082 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl22 | NM_001277113.1 | 4 | 946 | NP_001264042.1 |
| NM_001277114.1 | 4 | 1016 | NP_001264043.1 | |
| NM_009079.3 | 4 | 996 | NP_033105.1 | |
| NR_102291.1 | 5 | 1150 | ||
| NR_102292.1 | 5 | 1146 | ||
| AK009094.1 | 4 | 863 | ||
| AK012339.1 | 1 | 130 | ||
| AK014372.1 | 1 | 130 | ||
| AK169078.1 | 4 | 849 | BAE40863.1 | |
| AK169731.1 | 4 | 862 | ||
| BC021344.1 | 4 | 855 | AAH21344.1 |
More Information
Set Membership: |
|

Genomic Map