Assay Details
Target Gene Details
Entrez Gene ID: | 212190 |
Gene Name: | UBX domain protein 10 |
Gene Aliases: |
5730509E04Rik, A830047D02, Ubxd3 |
Location: |
Chr.4:138709837-138738438 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubxn10 | NM_001285928.1 | 4 | 1885 | NP_001272857.1 |
NM_001285929.1 | 3 | 1774 | NP_001272858.1 | |
NM_001285930.1 | 3 | 1638 | NP_001272859.1 | |
NM_178671.5 | 2 | 1507 | NP_848786.1 | |
NR_104373.1 | 2 | 1507 | ||
AK045964.1 | 2 | 1465 | BAC32548.1 | |
AK046949.1 | 4 | 1886 | BAC32924.1 | |
AK077656.1 | 2 | 1509 | BAC36931.1 | |
AK080662.1 | 2 | 1473 | BAC37971.1 | |
AK133185.1 | 2 | 1493 | BAE21549.1 | |
AK161245.1 | 2 | 1494 | BAE36264.1 | |
AK161296.1 | 2 | 1470 | BAE36303.1 | |
BC027099.1 | 3 | 1638 | AAH27099.1 |
