Assay Details
Target Gene Details
Entrez Gene ID: | 65105 |
Gene Name: | ADP-ribosylation factor-like 6 interacting protein 4 |
Gene Aliases: |
AA408210, AA408365, Aip-4, Srp25 |
Location: |
Chr.5:124116108-124118195 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl6ip4 | NM_144509.2 | 2 | 732 | NP_653092.1 |
| AB035383.1 | 2 | 732 | BAA94743.1 | |
| AK133934.1 | 2 | 548 | BAE21935.1 | |
| BC063748.1 | 2 | 699 | AAH63748.1 |

Genomic Map